missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimetridazole, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Dimetridazole
Specifications
Specifications
| Chemical Name or Material | Dimetridazole |
| CAS | 551-92-8 |
| Analyte or Component Names | Dimetridazole |
| Shipping Condition | Room Temperature |
| Molecular Formula | C5 H7 N3 O2 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Dimetridazole, 1H-Imidazole, 1,2-dimethyl-5-nitro-, Imidazole, 1,2-dimethyl-5-nitro- (7CI,8CI), 1,2-Dimethyl-5-nitro-1H-imidazole, 1,2-Dimethyl-5-nitroimidazole, 5-Nitro-1,2-dimethylimidazole, 8595RP, Dimetridazol, Dimetridazole, Dimetrizadole, Dimetronidazole, Emtryl, Emtrylvet, Emtrymix, NSC 226253 |
| InChI Formula | InChI=1S/C5H7N3O2/c1-4-6-3-5(7(4)2)8(9)10/h3H,1-2H3 |
| SMILES | Cc1ncc([N+](=O)[O-])n1C |
| IUPAC Name | 1,2-dimethyl-5-nitroimidazole |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction