missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimetindene N-Oxide Dihydrochloride, Mikromol™
Description
Dimetindene N-Oxide Dihydrochloride
Specifications
Specifications
| Chemical Name or Material | Dimetindene N-Oxide Dihydrochloride |
| CAS | 2724726-98-9 |
| Analyte or Component Names | Dimetindene N-Oxide Dihydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H24 N2 O . 2 Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1H-Indene-2-ethanamine, N,N-dimethyl-3-[1-(2-pyridinyl)ethyl]-, N-oxide, dihydrochloride, Dimetindene N-Oxide Dihydrochloride |
| InChI Formula | InChI=1S/C20H24N2O.2ClH/c1-15(19-10-6-7-12-21-19)20-17(11-13-22(2,3)23)14-16-8-4-5-9-18(16)20;;/h4-10,12,15H,11,13-14H2,1-3H3;2*1H |
| SMILES | Cl.Cl.CC(C1=C(CC[N+](C)(C)[O-])Cc2ccccc12)c3ccccn3 |
| IUPAC Name | N,N-dimethyl-2-[3-(1-pyridin-2-ylethyl)-1H-inden-2-yl]ethanamine oxide;dihydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?