missing translation for 'onlineSavingsMsg'
Learn More

Dimethyltin Chloride, TRC

Product Code. 30387036
Change view
Click to view available options
Quantity:
1 g
5 g
Product Code. Quantity
30387036 5 g
30370928 1 g
2 options
This item is not returnable. View return policy

Product Code. 30387036

Brand: Toronto Research Chemicals TRCD4800305G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 753-73-1
Formula Weight 219.8869
InChI Formula InChI=1S/2CH3.2ClH.Sn/h2*1H3;2*1H;/q;;;;+2/p-2
IUPAC Name dichloro(dimethyl)stannane
Molecular Formula C2 H6 Cl2 Sn
Molecular Weight (g/mol) 219.68
SMILES C[Sn](C)(Cl)Cl
Synonym Dimethyltin Dichloride, Stannane, dichlorodimethyl-, Tin, dimethyl-, dichloride (6CI), Dichlorodimethylstannane, Cotin 210, Dichlorodimethyltin, Dimethyldichlorostannane, Dimethyldichlorotin, Dimethylstannyl dichloride, Dimethyltin dichloride, Dimethyltin(IV) dichloride, NSC 55159, Nesstin P
Chemical Name or Material Dimethyltin-dichloride

Specifications

Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.