missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethylthiocarbamoyl Chloride, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Dimethylthiocarbamoyl Chloride |
| CAS | 16420-13-6 |
| Molecular Formula | C3H6ClNS |
| Quantity | 5 g |
| Synonym | N,N-Dimethylcarbamothioic Chloride, Dimethylthiocarbamyl Chloride, 1-Chloro-N,N-dimethylthioformamide, Dimethylcarbamothioic Chloride, Dimethylcarbamothioyl Chloride, N,N-Dimethylcarbamothioyl Chloride, N,N-Dimethylchlorothiocarbamate, N,N-Dimethylthiocarbamic Acid Chloride, N,N-Dimethylthiocarbamoyl Chloride, N,N-Dimethylthiocarbamyl Chloride, N,N'-Dimethylchlorothioformamide |
| InChI Formula | InChI=1S/C3H6ClNS/c1-5(2)3(4)6/h1-2H3 |
| SMILES | CN(C)C(=S)Cl |
| IUPAC Name | N,N-dimethylcarbamothioyl chloride |
| Molecular Weight (g/mol) | 123.6 |
| Formula Weight | 122.9909 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction