missing translation for 'onlineSavingsMsg'
Learn More

Dimethylsuberimidate dihydrochloride, Thermo Scientific Chemicals

Catalog Number p-7058773
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
34490-86-3
C10H22Cl2N2O2
273.198
MFCD00012574
ILKCDNKCNSNFMP-UHFFFAOYSA-N
dimethyl suberimidate dihydrochloride, octanediimidic acid, dimethyl ester, dihydrochloride, dimethyl octanebis imidate dihydrochloride, suberimidic acid dimethyl ester dihydrochloride, octanediimidic acid, 1,8-dimethyl ester, hydrochloride 1:2, 1,8-dimethyl octanediimidate dihydrochloride, suberimidic acid, dimethyl ester, dihydrochloride, acmc-1aiyg, 1,8-dimethoxyoctane-1,8-diyldiammonium dichloride, dimethylsuberimidate dihydrochloride
118696
dimethyl octanediimidate;dihydrochloride
COC(=N)CCCCCCC(=N)OC.Cl.Cl
This item is not returnable. View return policy
34490-86-3
C10H22Cl2N2O2
273.198
MFCD00012574
ILKCDNKCNSNFMP-UHFFFAOYSA-N
dimethyl suberimidate dihydrochloride, octanediimidic acid, dimethyl ester, dihydrochloride, dimethyl octanebis imidate dihydrochloride, suberimidic acid dimethyl ester dihydrochloride, octanediimidic acid, 1,8-dimethyl ester, hydrochloride 1:2, 1,8-dimethyl octanediimidate dihydrochloride, suberimidic acid, dimethyl ester, dihydrochloride, acmc-1aiyg, 1,8-dimethoxyoctane-1,8-diyldiammonium dichloride, dimethylsuberimidate dihydrochloride
118696
dimethyl octanediimidate;dihydrochloride
COC(=N)CCCCCCC(=N)OC.Cl.Cl

Crosslinking of proteins and poly(allylamine)

Dimethylsuberimidate dihydrochloride is used as a homobifunctional cross-linking reagent for amino groups. It is also used for the modification of tyrosinase to produce biocatalytic enzymatic flow-through detectors. It serves as a cross-linker for preparation of collagen-calcium phosphate composites. Further, it is used in the treatment of avian myeloblastosis virus.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 213°C to 214°C
Quantity 2 g
Sensitivity Moisture sensitive
Solubility Information Soluble in water.
Formula Weight 273.2
Physical Form Crystalline
Chemical Name or Material Dimethylsuberimidate dihydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'