missing translation for 'onlineSavingsMsg'
Learn More

Dimethylsilyldiethylamine, 95%

Product Code. 11342186
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11342186 25 g 25g
11332186 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11342186 Supplier Thermo Scientific Alfa Aesar Supplier No. L03503.14
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13686-66-3 | C6H16NSi | 130.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13686-66-3
Molecular Formula C6H16NSi
Molecular Weight (g/mol) 130.29
MDL Number MFCD00048523
InChI Key ADTGAVILDBXARD-UHFFFAOYSA-N
Synonym dimethylsilyldiethylamine, n,n-diethyl-1,1-dimethylsilylamine, diethylamino dimethylsilyl, silanamine, n,n-diethyl-1,1-dimethyl, silanamine,n,n-diethyl-1,1-dimethyl, diethylamino dimethyl silicon, diethylaminodimethylsilane, n,n-diethyl-1,1-dimethylsilanamine
PubChem CID 6335300
IUPAC Name (diethylamino)dimethylsilyl
SMILES CCN(CC)[Si](C)C

Specifications

Boiling Point 109°C to 110°C
Quantity 25 g
Flash Point <−5°C (23°F)
Refractive Index 1.408
Sensitivity Moisture sensitive
Density 0.8
UN Number UN1993
Beilstein 741862
Formula Weight 131.3
Percent Purity 95%
Chemical Name or Material Dimethylsilyldiethylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.