missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethylphosphoramidous Dichloride, 97%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
Packaging:
Glass bottle
Unit Size:
5mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless to Yellow |
| Density | 1.2710g/mL |
| Boiling Point | 150.0°C |
| Flash Point | −23°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97% |
| Packaging | Glass bottle |
| Linear Formula | (CH3)2NPCl2 |
| Refractive Index | 1.5040 to 1.5060 (20°C, 589nm) |
| Quantity | 5 mL |
| Show More |
Safety and Handling
Product Identifier
- Dimethylphosphoramidous dichloride
- Danger
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- H225-Highly flammable liquid and vapour.
- EUH014-Reacts violently with water.
- EUH029-Contact with water liberates toxic gas.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction