missing translation for 'onlineSavingsMsg'
Learn More

Dimethylphosphinic chloride, 97+%

Product Code. 11357248
Change view
Click to view available options
Quantity:
0.5 g
2 g
Unit Size:
0.5g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357248 2 g 2g
11347248 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357248 Supplier Thermo Scientific Alfa Aesar Supplier No. 030292.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1111-92-8 | C2H6ClOP | 112.493 g/mol

Dimethylphosphinic chloride is used in the protection of amino acids in peptide synthesis. It initiates the luminescence of ABI-NH2. It is utilized in the reactivity of organophosphate-based nerve gases such as sarin gas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1111-92-8
Molecular Formula C2H6ClOP
Molecular Weight (g/mol) 112.493
MDL Number MFCD00002078
InChI Key CVNMBKFJYRAHPO-UHFFFAOYSA-N
Synonym dimethylphosphinic chloride, phosphinic chloride, dimethyl, dimethylphosphinoyl chloride, phosphinic chloride,dimethyl-6ci,7ci,8ci,9ci, me2p o cl, dimethylphosphinoylchloride, dimethylphosphinyl chloride, acmc-1bo3s, dimethylphosphin1c chloride, dimethyl phosphinyl chloride
PubChem CID 517954
IUPAC Name [chloro(methyl)phosphoryl]methane
SMILES CP(=O)(C)Cl

Specifications

Melting Point 68°C
Boiling Point 204°C
Quantity 2 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 112.51
Percent Purity ≥97%
Physical Form Crystalline
Chemical Name or Material Dimethylphosphinic chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.