missing translation for 'onlineSavingsMsg'
Learn More

Dimethylphosphinic chloride, 97+%

Product Code. p-7049851
Click to view available options
Quantity:
0.5 g
2 g
Unit Size:
0.5g
2g
This item is not returnable. View return policy

Product Code. 11347248

Brand: Thermo Scientific Alfa Aesar 030292.02

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1111-92-8 | C2H6ClOP | 112.493 g/mol

Dimethylphosphinic chloride is used in the protection of amino acids in peptide synthesis. It initiates the luminescence of ABI-NH2. It is utilized in the reactivity of organophosphate-based nerve gases such as sarin gas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1111-92-8
Molecular Formula C2H6ClOP
Molecular Weight (g/mol) 112.493
MDL Number MFCD00002078
InChI Key CVNMBKFJYRAHPO-UHFFFAOYSA-N
Synonym dimethylphosphinic chloride, phosphinic chloride, dimethyl, dimethylphosphinoyl chloride, phosphinic chloride,dimethyl-6ci,7ci,8ci,9ci, me2p o cl, dimethylphosphinoylchloride, dimethylphosphinyl chloride, acmc-1bo3s, dimethylphosphin1c chloride, dimethyl phosphinyl chloride
PubChem CID 517954
IUPAC Name [chloro(methyl)phosphoryl]methane
SMILES CP(=O)(C)Cl

Specifications

Melting Point 68°C
Boiling Point 204°C
Quantity 0.5 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 112.51
Percent Purity ≥97%
Physical Form Crystalline
Chemical Name or Material Dimethylphosphinic chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.