missing translation for 'onlineSavingsMsg'
Learn More

Dimethylphenylsilane, 98%, ACROS Organics™

Product Code. 11945821 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
25g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11945821 25g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11945821 Supplier Thermo Scientific Acros Supplier No. 215930250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Reduction reagent
TRUSTED_SUSTAINABILITY

Specifications

CAS 766-77-8
Color Colorless
Density 0.8890g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C8H11Si
Linear Formula C6H5SiH(CH3)2
Refractive Index 1.4970 to 1.4990 (20°C, 589nm)
Quantity 25 g
Synonym dimethylphenylsilane, dimethyl phenyl silane, phenyldimethylsilane, silane, dimethylphenyl, benzene, dimethylsilyl, dimethylphenyl silane, c8h12si, dimethyl phenyl silyl, dimethyl-phenylsilicon
InChI Key OIKHZBFJHONJJB-UHFFFAOYSA-N
SMILES C[Si](C)C1=CC=CC=C1
IUPAC Name dimethyl(phenyl)silicon
Molecular Weight (g/mol) 135.261
PubChem CID 6327656
Formula Weight 136.27
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material Dimethylphenylsilane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.