missing translation for 'onlineSavingsMsg'
Learn More

Dimethylphenylsilane, 97%

Product Code. p-7044099
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11391617 10 g 10g
11301627 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11391617 Supplier Thermo Scientific Alfa Aesar Supplier No. L04558.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 766-77-8 | C8H11Si | 135.261 g/mol

Dimethylphenylsilane is a reagent for enol ether synthesis. It acts as a catalyst. Also used as a precursor in Optical Emission Spectroscopy of Plasma Deposition Processes. It can react with vinylbenzene to produce (b-Phenyl-ethyl)-dimethylphenyl-silan.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethylphenylsilane is a reagent for enol ether synthesis. It acts as a catalyst. Also used as a precursor in Optical Emission Spectroscopy of Plasma Deposition Processes. It can react with vinylbenzene to produce (b-Phenyl-ethyl)-dimethylphenyl-silan.

Solubility
Not miscible in water.

Notes
Moisture sensitive. Incompatible materials are Strong oxidizing agents, Strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 766-77-8
Molecular Formula C8H11Si
Molecular Weight (g/mol) 135.261
MDL Number MFCD00008257
InChI Key OIKHZBFJHONJJB-UHFFFAOYSA-N
Synonym dimethylphenylsilane, dimethyl phenyl silane, phenyldimethylsilane, silane, dimethylphenyl, benzene, dimethylsilyl, dimethylphenyl silane, c8h12si, dimethyl phenyl silyl, dimethyl-phenylsilicon
PubChem CID 6327656
IUPAC Name dimethyl(phenyl)silicon
SMILES C[Si](C)C1=CC=CC=C1

Specifications

Boiling Point 156°C to 157°C
Melting Point -124°C
Quantity 10 g
UN Number UN1993
Beilstein 2204906
Flash Point 35°C (95°F)
Solubility Information Not miscible in water.
Formula Weight 136.27
Percent Purity 97%
Refractive Index 1.497
Density 0.879
Chemical Name or Material Dimethylphenylsilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.