missing translation for 'onlineSavingsMsg'
Learn More

Dimethylphenylphosphine, 96%

Product Code. 11489106
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11489106 5 g 5g
11479106 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11489106 Supplier Thermo Scientific Alfa Aesar Supplier No. A17975.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 672-66-2 | C8H11P | 138.15 g/mol

Dimethylphenylphosphine is used as a ligand in transition metal complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethylphenylphosphine is used as a ligand in transition metal complexes.

Solubility
Soluble in organic solvents.

Notes
Air sensitive. Store away from oxidizing agents, air, acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 672-66-2
Molecular Formula C8H11P
Molecular Weight (g/mol) 138.15
MDL Number MFCD00008509
InChI Key HASCQPSFPAKVEK-UHFFFAOYSA-N
Synonym dimethylphenylphosphine, dimethyl phenyl phosphine, dimethyl phenyl phosphane, me2pph, phenyldimethylphosphine, pme2ph, phosphine, dimethylphenyl, pubchem6420, dimethyl phenylphosphine
PubChem CID 69597
ChEBI CHEBI:30671
IUPAC Name dimethyl(phenyl)phosphane
SMILES CP(C)C1=CC=CC=C1

Specifications

Density 0.971
Boiling Point 75°C to 79°C (12 mmHg)
Flash Point 50°C (122°F)
Odor Unpleasant
Refractive Index 1.563
Quantity 5 g
UN Number UN1993
Beilstein 742064
Sensitivity Air Sensitive
Solubility Information Soluble in organic solvents.
Formula Weight 138.15
Percent Purity 97%
Chemical Name or Material Dimethylphenylphosphine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.