missing translation for 'onlineSavingsMsg'
Learn More

Dimethyloctadecyl[3-(trimethoxysilyl)propyl]ammonium chloride, 60% in methanol

Product Code. 10153111 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10153111 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10153111 Supplier Thermo Scientific Acros Supplier No. 338531000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 27668-52-6 | C26H58ClNO3Si | 496.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Specifications

CAS 27668-52-6
Color Yellow
Density 0.8900g/mL
Flash Point 15°C
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C26H58ClNO3Si
Linear Formula [(CH3O)3Si(CH2)3N(CH3)2(CH2)17CH3]Cl
MDL Number MFCD00054224
Quantity 100 mL
Specific Gravity 0.89
Synonym quat-silsesquioxane, dmoap, dow corning 5700, caswell no. 892b, dimethyloctadecyl 3-trimethoxysilyl propyl ammonium chloride, unii-iq36o85wq4, n,n-dimethyl-n-3-trimethoxysilyl propyl octadecan-1-aminium chloride, epa pesticide chemical code 107401, 3-trimethoxysilyl propyl dimethyl octadecyl ammonium chloride, 3-trimethoxysilyl propyldimethyloctadecylammonium
Solubility Information Solubility in water: reacts
InChI Key WSFMFXQNYPNYGG-UHFFFAOYSA-M
SMILES [Cl-].CCCCCCCCCCCCCCCCCC[N+](C)(C)CCC[Si](OC)(OC)OC
IUPAC Name dimethyl-octadecyl-(3-trimethoxysilylpropyl)azanium;chloride
Molecular Weight (g/mol) 496.29
PubChem CID 62827
Formula Weight 496.29
Percent Purity 60% in methanol
Physical Form Solution
Chemical Name or Material Dimethyloctadecyl[3-(trimethoxysilyl)propyl]ammonium chloride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dimethyloctadecyl¢3-(trimethoxysilyl)propyl!ammonium chloride, 60% in methanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H370-Causes damage to organs.
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.