missing translation for 'onlineSavingsMsg'
Learn More

Dimethylglyoxime Sodium Salt, Pure, Fisher Chemical

Product Code. 10417830 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 g
Packaging:
Plastic powder jar
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10417830 250 g Plastic powder jar 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10417830 Supplier Fisher Chemical Supplier No. D/3950/50

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 75006-64-3
Color White
Physical Form Powder/Solid
Quantity 250 g
Molecular Formula C4H22N2Na2O10
MDL Number MFCD00150288
Synonym dimethylglyoxime disodium salt octahydrate, c4h6n2o2.2na.8h2o, sodium dimethylglyoxime octahydrate, dimethylglyoxime di-sodium salt octahydrate
InChI Key IJJDRGUPRATWQX-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.O.O.[Na+].[Na+].CC(=N[O-])C(C)=N[O-]
IUPAC Name disodium oxido[3-(oxidoimino)butan-2-ylidene]amine octahydrate
Molecular Weight (g/mol) 304.20
PubChem CID 71310106
Formula Weight 304.19g/mol
Packaging Plastic powder jar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.