missing translation for 'onlineSavingsMsg'
Learn More

Dimethylfraxetin, MedChemExpress

Product Code. 16441298
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16441298

Brand: Medchem Express HYN0085

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dimethylfraxetin is a Carbonic anhydrase inhibitor, with a Ki value of 0.0097 μM.

Dimethylfraxetin is a Carbonic anhydrase inhibitor, with a Ki value of 0.0097 μM.

Specifications

Chemical Name or Material Dimethylfraxetin
CAS 6035-49-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C12H12O5
Quantity 5 mg
Synonym 6,7,8-Trimethoxycoumarin Fraxetin dimethyl ether
Solubility Information DMSO : 125 mg/mL (529.17 mM; Need ultrasonic and warming)
SMILES O=C1C=CC2=CC(OC)=C(OC)C(OC)=C2O1
Molecular Weight (g/mol) 236.22
Formula Weight 236.22
Percent Purity 99.81%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.