missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DimethylFormamide, Extra Pure, SLR, Fisher Chemical™
45.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
45.58 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "22914" to get your promotional price.
45.58 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
missing translation for 'orderingAttributeHoverText'
Quantity:
500 mL
1 L
2.5 L
5 L
25 L
Packaging:
Amber Glass Bottle
HDPE Bottle
Plastic Container
Description
DimethylFormamide, Extra Pure, SLR
Spécification
pH | 6 to 8 |
Melting Point | -61°C |
Color | Colorless |
Boiling Point | 153°C |
Quantity | 2.5 L |
Packaging | HDPE Bottle |
Physical Form | Liquid |
Vapor Pressure | 4.9mbar at 20°C |
Viscosity | 0.8 mPaS at 20°C |
Molecular Weight (g/mol) | 73.10 |
Afficher plus |
Sécurité et traitement
Identifiant produit
- Dimethylformamide
- Danger
- Flammable liquid Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
- H226-Flammable liquid and vapour.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H360D-May damage the unborn child.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'