missing translation for 'onlineSavingsMsg'
Learn More

Dimethylethylhexadecylammonium bromide, 99%

Product Code. 10144790 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10144790 500 g Glass bottle 500g
10753021 5 g Glass bottle 5g
10532041 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10144790 Supplier Thermo Scientific Acros Supplier No. 211875000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 124-03-8 | C20H44BrN | 378.483 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-03-8
Molecular Formula C20H44BrN
Molecular Weight (g/mol) 378.483
MDL Number MFCD00011774
InChI Key VUFOSBDICLTFMS-UHFFFAOYSA-M
Synonym Cetyldimethylethylammonium bromide
PubChem CID 31280
IUPAC Name ethyl-hexadecyl-dimethylazanium;bromide
SMILES CCCCCCCCCCCCCCCC[N+](C)(C)CC.[Br-]

Specifications

Color White
Physical Form Powder
pH 6 to 8 (1% soln.)
Quantity 500 g
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 99%
Linear Formula CH3(CH2)15N(CH3)2(C2H5)Br
Beilstein 04, III, 425
Merck Index 15, 2028
Formula Weight 378.48
Percent Purity ≥98.5%
Chemical Name or Material Dimethylethylhexadecylammonium bromide, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Dimethylethylhexadecylammonium bromide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.