Learn More
Dimethylethoxysilane, 94%, Thermo Scientific Chemicals
£18.20 - £182.00
Chemical Identifiers
CAS | 14857-34-2 |
---|---|
Molecular Formula | C4H12OSi |
Molecular Weight (g/mol) | 104.22 |
MDL Number | MFCD00026750 |
InChI Key | DRUOQOFQRYFQGB-UHFFFAOYSA-N |
Synonym | ethoxydimethylsilane, dimethylethoxysilane, silane, ethoxydimethyl, dimethyl ethoxysilane, ethoxy dimethyl silicon, dimethylethoxysilane, ammonia-free, ethoxydimethylsilyl, ethoxydimethyl-silan, sgqhhgcihcup@, 4-04-00-03991 beilstein handbook reference |
PubChem CID | 6328550 |
IUPAC Name | ethoxy(dimethyl)silicon |
SMILES | CCO[Si](C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10339353
|
Thermo Scientific
392000050 |
5 g | Glass bottle |
£18.20
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10762244
|
Thermo Scientific Acros
392000250 |
25 g | Glass bottle |
£52.70
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10772244
|
Thermo Scientific Acros
392001000 |
100 g | Glass bottle |
£182.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentChemical Identifiers
14857-34-2 | |
104.22 | |
DRUOQOFQRYFQGB-UHFFFAOYSA-N | |
6328550 | |
CCO[Si](C)C |
C4H12OSi | |
MFCD00026750 | |
ethoxydimethylsilane, dimethylethoxysilane, silane, ethoxydimethyl, dimethyl ethoxysilane, ethoxy dimethyl silicon, dimethylethoxysilane, ammonia-free, ethoxydimethylsilyl, ethoxydimethyl-silan, sgqhhgcihcup@, 4-04-00-03991 beilstein handbook reference | |
ethoxy(dimethyl)silicon |
Specifications
14857-34-2 | |
54.0°C | |
Authentic | |
C4H12OSi | |
1.3670 to 1.3690 (20°C, 589nm) | |
ethoxydimethylsilane, dimethylethoxysilane, silane, ethoxydimethyl, dimethyl ethoxysilane, ethoxy dimethyl silicon, dimethylethoxysilane, ammonia-free, ethoxydimethylsilyl, ethoxydimethyl-silan, sgqhhgcihcup@, 4-04-00-03991 beilstein handbook reference | |
DRUOQOFQRYFQGB-UHFFFAOYSA-N | |
ethoxy(dimethyl)silicon | |
6328550 | |
94% | |
Dimethylethoxysilane |
Colorless | |
−5°C | |
92% min. (GC) | |
CH3CH2OSiH(CH3)2 | |
MFCD00026750 | |
Solubility in water: reacts | |
CCO[Si](C)C | |
104.22 | |
104.22 | |
Liquid |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 238-921-7
- Dimethylethoxysilane
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only