missing translation for 'onlineSavingsMsg'
Learn More

Dimethylethoxysilane, 94%

Product Code. 11329185
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11329185 100 g 100g
11319185 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11329185 Supplier Thermo Scientific Alfa Aesar Supplier No. L16476.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14857-34-2 | C4H11OSi | 103.216 g/mol

Dimethylethoxysilane (DMES), a volatile liquid, is used by NASA to waterproof the heatprotective silica tiles and blankets on the space shuttle. DMES readily produces a variety of products in the presence of small amounts of moisture or bases. DMES readily produces a variety of products in the presence of small amounts of moisture or bases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethylethoxysilane (DMES), a volatile liquid, is used by NASA to waterproof the heatprotective silica tiles and blankets on the space shuttle. DMES readily produces a variety of products in the presence of small amounts of moisture or bases. DMES readily produces a variety of products in the presence of small amounts of moisture or bases.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture, water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14857-34-2
Molecular Formula C4H11OSi
Molecular Weight (g/mol) 103.216
MDL Number MFCD00026750
InChI Key DRUOQOFQRYFQGB-UHFFFAOYSA-N
Synonym ethoxydimethylsilane, dimethylethoxysilane, silane, ethoxydimethyl, dimethyl ethoxysilane, ethoxy dimethyl silicon, dimethylethoxysilane, ammonia-free, ethoxydimethylsilyl, ethoxydimethyl-silan, sgqhhgcihcup@, 4-04-00-03991 beilstein handbook reference
PubChem CID 6328550
IUPAC Name ethoxy(dimethyl)silicon
SMILES CCO[Si](C)C

Specifications

Boiling Point 55°C
Quantity 100 g
Flash Point 15°C (59°F)
Refractive Index 1.368
Sensitivity Moisture sensitive
Density 0.757
UN Number UN1993
Beilstein 1731481
Solubility Information Hydrolyzes in water.
Formula Weight 104.23
Percent Purity 94%
Chemical Name or Material Dimethylethoxysilane, ammonia-free
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.