missing translation for 'onlineSavingsMsg'
Learn More

Dimethylepyridine-2,5-dicarboxylate, 97%, Thermo Scientific™

Product Code. 10203522
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10203522 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10203522 Supplier Thermo Scientific Maybridge Supplier No. RF02989EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 881-86-7
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C9H9NO4
MDL Number MFCD00034767
Quantity 10 g
Synonym dimethyl 2,5-pyridinedicarboxylate, 2,5-pyridinedicarboxylic acid, dimethyl ester, dimethylpyridine-2,5-dicarboxylate, 2,5-dimethyl pyridine-2,5-dicarboxylate, dimethyl isocinchomeronate, pyridine-2,5-dicarboxylic acid dimethyl ester, dimethyl 2,5-pyridine dicarboxylate, acmc-1bkgx, maybridge1_006391, methyl 6-methoxycarbonylnicotinate
InChI Key TUGSJNQAIMFEDY-UHFFFAOYSA-N
SMILES COC(=O)C1=CN=C(C=C1)C(=O)OC
IUPAC Name dimethyl pyridine-2,5-dicarboxylate
Molecular Weight (g/mol) 195.174
PubChem CID 234911
Formula Weight 195.17
Percent Purity 97%
Chemical Name or Material Dimethylepyridine-2, 5-dicarboxylate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.