missing translation for 'onlineSavingsMsg'
Learn More

(+)-Dimethyl L-tartrate, 98+%

Product Code. 10429470 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10429470 100 g Glass bottle 100g
10522391 25 g Glass bottle 25g
10016093 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10429470 Supplier Thermo Scientific Acros Supplier No. 172491000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 608-68-4 | C6H10O6 | 178.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 608-68-4
Molecular Formula C6H10O6
Molecular Weight (g/mol) 178.14
MDL Number MFCD00064437
InChI Key PVRATXCXJDHJJN-QWWZWVQMSA-N
Synonym +-dimethyl l-tartrate, dimethyl l-tartrate, 2r,3r-dimethyl 2,3-dihydroxysuccinate, dimethyl +-tartrate, l-+-tartaric acid dimethyl ester, dimethyl l-+-tartrate, 1,4-dimethyl 2r,3r-2,3-dihydroxybutanedioate, butanedioic acid, 2,3-dihydroxy-2r,3r-, 1,4-dimethyl ester, dimethyl 2r,3r-2,3-dihydroxybutanedioate, l-tartaric acid, dimethyl ester
PubChem CID 11851
IUPAC Name dimethyl (2R,3R)-2,3-dihydroxybutanedioate
SMILES COC(=O)C(C(C(=O)OC)O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 48°C to 50°C
Color White
Boiling Point 157°C to 159°C (15.0 hPa)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98+% min. (GC)
Packaging Glass bottle
Linear Formula [-CH(OH)CO2CH3]2
Quantity 100 g
Beilstein 03, 510
Fieser 13,124
Specific Rotation Condition + 21.00 (20.00°C c=10,H2O)
Specific Rotation + 21
Solubility Information Solubility in water: soluble in water
Formula Weight 178.14
Percent Purity 98%
Physical Form Crystals
Chemical Name or Material (+)-Dimethyl L-tartrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.