missing translation for 'onlineSavingsMsg'
Learn More

Dimethyle4-(4-methoxyphenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate, 97%, Thermo Scientific™

Catalog Number 10374102
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
73257-47-3
C18H21NO5
331.368
MFCD00085027
IAXDEFZXLVTHLU-UHFFFAOYSA-N
dimethyl 4-4-methoxyphenyl-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate, 3,5-dimethyl 4-4-methoxyphenyl-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate, dimethyl 4-4-methoxyphenyl-2,6-dimethyl-1,4-dihydro-3,5-pyridinedicarboxylate, maybridge1_007478, cbmicro_033161, 1,4-dihydro-2,6-dimethyl-4-4-methoxyphenyl-3,5-pyridinedicarboxylic acid dimethyl ester, dimethyl 1,4-dihydro-4-4-methoxyphenyl-2,6-dimethylpyridine-3,5-dicarboxylate, dimethyl 4-4-methoxyphenyl-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxy late, 3,5-pyridinedicarboxylic acid, 1,4-dihydro-4-4-methoxyphenyl-2,6-dimethyl-, dimethyl ester, 3,5-pyridinedicarboxylicacid, 1,4-dihydro-4-4-methoxyphenyl-2,6-dimethyl-, 3,5-dimethyl ester
614332
dimethyl 4-(4-methoxyphenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
CC1=C(C(C(=C(N1)C)C(=O)OC)C2=CC=C(C=C2)OC)C(=O)OC
This item is not returnable. View return policy
73257-47-3
C18H21NO5
331.368
MFCD00085027
IAXDEFZXLVTHLU-UHFFFAOYSA-N
dimethyl 4-4-methoxyphenyl-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate, 3,5-dimethyl 4-4-methoxyphenyl-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate, dimethyl 4-4-methoxyphenyl-2,6-dimethyl-1,4-dihydro-3,5-pyridinedicarboxylate, maybridge1_007478, cbmicro_033161, 1,4-dihydro-2,6-dimethyl-4-4-methoxyphenyl-3,5-pyridinedicarboxylic acid dimethyl ester, dimethyl 1,4-dihydro-4-4-methoxyphenyl-2,6-dimethylpyridine-3,5-dicarboxylate, dimethyl 4-4-methoxyphenyl-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxy late, 3,5-pyridinedicarboxylic acid, 1,4-dihydro-4-4-methoxyphenyl-2,6-dimethyl-, dimethyl ester, 3,5-pyridinedicarboxylicacid, 1,4-dihydro-4-4-methoxyphenyl-2,6-dimethyl-, 3,5-dimethyl ester
614332
dimethyl 4-(4-methoxyphenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
CC1=C(C(C(=C(N1)C)C(=O)OC)C2=CC=C(C=C2)OC)C(=O)OC
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 331.37
Percent Purity 97%
Chemical Name or Material Dimethyl 4-(4-methoxyphenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'