missing translation for 'onlineSavingsMsg'
Learn More

Dimethyldodecylamine, Mikromol™

Product Code. 17980646
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17980646 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17980646 Supplier Mikromol Supplier No. MM0451.05
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Dimethyldodecylamine
TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material Dimethyldodecylamine
CAS 112-18-5
Analyte or Component Names Dimethyldodecylamine
Shipping Condition Room Temperature
Molecular Formula C14 H31 N
Certifications/Compliance ISO 9001
Synonym Dimethyldodecylamine
InChI Formula InChI=1S/C14H31N/c1-4-5-6-7-8-9-10-11-12-13-14-15(2)3/h4-14H2,1-3H3
SMILES CCCCCCCCCCCCN(C)C
IUPAC Name N,N-dimethyldodecan-1-amine
Molecular Weight (g/mol) 213.4
Quantity 100 mg
Formula Weight 213.2456 g/mol
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.