missing translation for 'onlineSavingsMsg'
Learn More

Dimethyldistearylammonium Bromide, TRC

Product Code. 30381125
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381125 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30381125 Supplier Toronto Research Chemicals Supplier No. TRCD481195250MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dimethyldistearylammonium Bromide
CAS 3700-67-2
Molecular Formula C38H80BrN
Quantity 250 mg
Synonym 1-Octadecanaminium, N,N-dimethyl-N-octadecyl-, bromide (9CI), Ammonium, dimethyldioctadecyl-, bromide (8CI), Dimethyldioctadecylammonium bromide (6CI,7CI), Bis(octadecyl)dimethylammonium bromide, DDAB, DODA(Br), DODAB, DSDMAB, Di-n-octadecyldimethylammonium bromide, Dimethyldioctadecylammomium bromide, Dioctadecanyldimethylammonium bromide, Dioctadecyldimethylammonium bromide, Distearyldimethylammonium bromide, GERBU Adjuvant 10, GERBU Adjuvant 100, N,N-Distearyl-N,N-dimethylammonium bromide, VSA 3
InChI Formula InChI=1S/C38H80N.BrH/c1-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-37-39(3,4)38-36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-2;/h5-38H2,1-4H3;1H/q+1;/p-1
SMILES [Br-].CCCCCCCCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCCCCCCCC
Molecular Weight (g/mol) 630.95
Formula Weight 629.5474
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.