missing translation for 'onlineSavingsMsg'
Learn More

Dimethyldiphenyltin

Product Code. 11374557
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11374557 5 g 5g
11364557 1 g 1g
11384557 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11374557 Supplier Thermo Scientific Alfa Aesar Supplier No. 039617.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1080-43-9 | C14H16Sn | 302.992 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1080-43-9
Molecular Formula C14H16Sn
Molecular Weight (g/mol) 302.992
MDL Number MFCD00058812
InChI Key INCQQSKGFIBXAY-UHFFFAOYSA-N
Synonym dimethyldiphenyltin, acmc-1bqk3, stannane, dimethyldiphenyl
PubChem CID 11130460
IUPAC Name dimethyl(diphenyl)stannane
SMILES C[Sn](C)(C1=CC=CC=C1)C2=CC=CC=C2

Specifications

Quantity 5 g
UN Number UN2788
Solubility Information Insoluble in water.
Formula Weight 302.97
Physical Form Liquid
Chemical Name or Material Dimethyldiphenyltin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.