missing translation for 'onlineSavingsMsg'
Learn More

Dimethyldioctadecylammonium Chloride (mixture of C16 to C18 chain lengths), TRC

Product Code. 30384909
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384909 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30384909 Supplier Toronto Research Chemicals Supplier No. TRCD4808481G
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dimethyldioctadecylammonium chloride
CAS 107-64-2
Molecular Formula C38 H80 N . Cl
Quantity 1 g
Synonym 1-Octadecanaminium, N,N-dimethyl-N-octadecyl-, chloride (9CI), Ammonium, dimethyldioctadecyl-, chloride (8CI), Dimethyldioctadecylammonium chloride (6CI,7CI), Adogen TA 100, Aerosurf TA 101, Aliquat 207, Ammoeng 130, Ammonyx 2200P100, Arosurf TA 100, Arosurf TA 101, Arquad 218-100, Arquad 218-100P, Arquad 218P75E, Arquad 2HP, Arquad 2HP-75, Arquad R 40, CA 3475, Cation DS, Cation DSV, Cedequat TD 75, D 1821, D 86P, DHAC, DODA(Cl), DODAC, Dehyquart DAM, Di-n-octadecyldimethylammonium chloride, Dimethyldioctadecanylammonium chloride, Dimethyldistearylammonium chloride, Dioctadecyldimethylammonium chloride, Distearic dimethyl ammonium chloride, Distearyldimethylammonium chloride, Distearyldimonium chloride, Genamin DSAC, Genamin DSAC 20, Genamin DSAP, HYQMD 1821C, KD 83, Kemamine Q 9702CLP, Lipoquad 2HP75, N,N-Dioctadecyl-N,N-dimethylammonium chloride, Nikkol CA 3475V, Pionin B 8811, Quartamin D 85P, Quartamin D 86, Quartamin D 86P, Quartamin DM 86P, Quaternium 5, Sokalan 9200, Surfroyal DSAC, Swanol CA 3475, TA 101, Talofloc, Varisoft 100, Varisoft TA 100
InChI Formula InChI=1S/C38H80N.ClH/c1-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-37-39(3,4)38-36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-2;/h5-38H2,1-4H3;1H/q+1;/p-1
SMILES [Cl-].CCCCCCCCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCCCCCCCC
IUPAC Name dimethyl(dioctadecyl)ammonium;chloride
Molecular Weight (g/mol) 586.5
Formula Weight 585.5979
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.