missing translation for 'onlineSavingsMsg'
Learn More

Dimethyldimyristylammonium Bromide, TRC

Product Code. 30385739
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385739 2.5 g 2.50g
30376895 250 mg -
30388518 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385739 Supplier Toronto Research Chemicals Supplier No. TRCD4812832.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68105-02-2
Molecular Formula C30H64BrN
Molecular Weight (g/mol) 518.74
Synonym 1-Tetradecanaminium, N,N-dimethyl-N-tetradecyl-, bromide (9CI), Di-n-tetradecyldimethylammonium bromide, Dimethyldimyristylammonium bromide, Dimethylditetradecylammonium bromide, Dimyristyldimethylammonium bromide, Ditetradecyldimethylammonium bromide, Dimethylditetradecylammonium Bromide, N,N-dimethyl-N-tetradecyl-1-Tetradecanaminium bromide, Di-n-tetradecyldimethylammonium bromide, Dimethylditetradecylammonium bromide, Dimyristyldimethylammonium bromide, Ditetradecyldimethylammonium bromide
IUPAC Name dimethyl-di(tetradecyl)azanium;bromide
SMILES [Br-].CCCCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCCCC

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C30H64N.BrH/c1-5-7-9-11-13-15-17-19-21-23-25-27-29-31(3,4)30-28-26-24-22-20-18-16-14-12-10-8-6-2;/h5-30H2,1-4H3;1H/q+1;/p-1
Formula Weight 517.4222
Chemical Name or Material Dimethyldimyristylammonium Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.