missing translation for 'onlineSavingsMsg'
Learn More

Dimethylcyanamide, TRC

Product Code. 30378104
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30378104 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30378104 Supplier Toronto Research Chemicals Supplier No. TRCD4715055G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dimethylcyanamide
CAS 1467-79-4
Molecular Formula C3H6N2
Quantity 5 g
Synonym Cyanodimethylamine, Dimethylaminocarbonitrile, N,N-Dimethylcyanamide, N,N-Dimethylcyanoamine, N-Cyano-N-methylmethanamine, N-Cyanodimethylamine, NSC 7765
InChI Formula InChI=1S/C3H6N2/c1-5(2)3-4/h1-2H3
SMILES CN(C)C#N
IUPAC Name dimethylcyanamide
Molecular Weight (g/mol) 70.09
Formula Weight 70.0531
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.