missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethylarginine Dimethylaminohydrolase 1/DDAH1 Rabbit anti-Human, Rat, Clone: 1Q5F6, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Dimethylarginine Dimethylaminohydrolase 1/DDAH1 Monoclonal antibody specifically detects Dimethylarginine Dimethylaminohydrolase 1/DDAH1 in Human, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Dimethylarginine Dimethylaminohydrolase 1/DDAH1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 1Q5F6 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:2000 |
| Formulation | PBS, 0.05% BSA, 50% glycerol, pH7.3 |
| Gene Alias | DDAH-1, DDAHdimethylargininase-1, DDAHI, Dimethylargininase-1, dimethylarginine dimethylaminohydrolase 1FLJ25539, EC 3.5.3.18, FLJ21264, N(G), N(G)-dimethylarginine dimethylaminohydrolase 1, NG, NG-dimethylarginine dimethylaminohydrolase |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to a sequence within amino acids 186-285 of human Dimethylarginine Dimethylaminohydrolase 1/DDAH1 (O94760). IAIGSSESAQKALKIMQQMSDHRYDKLTVPDDIAANCIYLNIPNKGHVLLHRTPEEYPESAKVYEKLKDHMLIPVSMSELEKVDGLLTCCSVLINKKVDS |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?