missing translation for 'onlineSavingsMsg'
Learn More

Dimethylarginine Dimethylaminohydrolase 1/DDAH1 Antibody [DyLight 405], Novus Biologicals™

Product Code. 30731131 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30731131 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30731131 Supplier Novus Biologicals Supplier No. AF6530E

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Dimethylarginine Dimethylaminohydrolase 1/DDAH1 Polyclonal antibody specifically detects Dimethylarginine Dimethylaminohydrolase 1/DDAH1 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dimethylarginine Dimethylaminohydrolase 1/DDAH1
Applications Immunohistochemistry
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias DDAH-1, DDAHdimethylargininase-1, DDAHI, Dimethylargininase-1, dimethylarginine dimethylaminohydrolase 1FLJ25539, EC 3.5.3.18, FLJ21264, N(G), N(G)-dimethylarginine dimethylaminohydrolase 1, NG, NG-dimethylarginine dimethylaminohydrolase
Host Species Sheep
Immunogen E. coli-derived recombinant human Dimethylarginine Dimethylaminohydrolase 1/DDAH1, Ala2-Ser285, Accession # O94760
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23576
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.