missing translation for 'onlineSavingsMsg'
Learn More

Dimethylaminoacetyl chloride hydrochloride, tech. 85%

Product Code. 15490847
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15490847 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15490847 Supplier Thermo Scientific Alfa Aesar Supplier No. L18475.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 60853-81-8 | C4H9Cl2NO | 158.02 g/mol

Dimethylaminoacetyl chloride hydrochloride is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethylaminoacetyl chloride hydrochloride is used as a pharmaceutical intermediate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to moisture. Store un
TRUSTED_SUSTAINABILITY

Specifications

CAS 60853-81-8
Molecular Formula C4H9Cl2NO
MDL Number MFCD02094025
Quantity 5 g
UN Number UN3261
Beilstein 3980151
Sensitivity Moisture sensitive
Synonym 2-dimethylamino acetyl chloride hydrochloride, dimethylaminoacetyl chloride hydrochloride, n,n-dimethylglycinyl chloride hydrochloride, acmc-1b50m, dimethylaminoacetyl chloride-hcl, dimethylamino-acetyl chloride hcl, dimethylaminoacetylchloride hydrochloride, n,n-dimethylglycyl chloride hydrochloride, dimethylamino-acetyl chloride hydrochloride, 2-dimethylaminoacetyl chloride hydrochloride
InChI Key MVAXDKDOPWPFML-UHFFFAOYSA-N
SMILES Cl.CN(C)CC(Cl)=O
IUPAC Name 2-(dimethylamino)acetyl chloride;hydrochloride
Molecular Weight (g/mol) 158.02
PubChem CID 13994991
Formula Weight 158.03
Percent Purity 85%
Grade Technical
Chemical Name or Material Dimethylaminoacetyl chloride hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.