missing translation for 'onlineSavingsMsg'
Learn More

Dimethylaminoacetaldehyde dimethylacetal, 97%

Product Code. 10239083 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10239083 5 g Glass bottle 5g
10688214 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10239083 Supplier Thermo Scientific Acros Supplier No. 390910050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 38711-20-5 | C6H15NO2 | 133.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38711-20-5
Molecular Formula C6H15NO2
Molecular Weight (g/mol) 133.19
MDL Number MFCD00010205
InChI Key HUYAEQCJNXODLQ-UHFFFAOYSA-N
Synonym ethanamine, 2,2-dimethoxy-n,n-dimethyl, dimethylamino acetaldehyde dimethyl acetal, dimethylaminoacetaldehyde dimethyl acetal, dimethylaminoacetaldehyde dimethylacetal, n-2,2-dimethoxyethyl dimethylamine, 2-dimethylamino acetaldehyde dimethylacetal, 2,2-dimethoxyethyl dimethylamine, 2,2-dimethoxyethyl dimethyl amine, 2,2-dimethoxy-n,n-dimethylethanamin, acmc-209j1l
PubChem CID 123476
IUPAC Name 2,2-dimethoxy-N,N-dimethylethanamine
SMILES COC(CN(C)C)OC

Specifications

Color Colorless to Yellow
Density 0.9000g/mL
Boiling Point 64°C to 66°C (12.0 mmHg)
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (CH3)2NCH2CH(OCH3)2
Refractive Index 1.4100 to 1.412
Quantity 5 g
Specific Gravity 0.9
Formula Weight 133.19
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Dimethylaminoacetaldehyde dimethylacetal
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dimethylaminoacetaldehyde dimethylacetal
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.