missing translation for 'onlineSavingsMsg'
Learn More

Dimethylamine Hydrochloride, TRC

Product Code. 30376813
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30376813 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30376813 Supplier Toronto Research Chemicals Supplier No. TRCD44673010G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Methylmethanamine Hydrochloride
CAS 506-59-2
Purity Grade Notes HPLC
Molecular Formula C2 H7 N . Cl H
Quantity 10 g
Synonym N-Methylmethanamine Hydrochloride, Dacarbazine Imp. D (EP) as Hydrochloride, Metformin Hydrochloride Imp. F (EP) as Hydrochloride, Imatinib Mesilate Imp. I (Pharmeuropa), Dimethylamine Hydrochloride
InChI Formula InChI=1S/C2H7N.ClH/c1-3-2;/h3H,1-2H3;1H
SMILES Cl.CNC
IUPAC Name N-methylmethanamine;hydrochloride
Molecular Weight (g/mol) 81.54
Formula Weight 81.0345
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.