missing translation for 'onlineSavingsMsg'
Learn More

Dimethylamine hydrochloride, 98+%, Thermo Scientific Chemicals

Product Code. 11400093
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11400093 5000 g 5000g
11490083 250 g 250g
11480083 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11400093 Supplier Thermo Scientific Alfa Aesar Supplier No. A12133.0I
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Dimethylamine hydrochloride is used as an intermediate in the manufacture of pharmaceuticals like ranitidine and metformin, tramadol and amlodipine. It is used as a precursor of atrazine. It is associated with sodium acetate and used to carry out the Willgerodt- Kindler reaction to prepare amides. Its free base reacts with carbon disulfide to get dimethyldithiocarbamate which is used in rubber vulcanization. It is involved in the synthesis of dimethyl-(1-methyl-pyrrol-2-ylmethyl)-amine by reacting with 1-methyl pyrrole and formaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethylamine hydrochloride is used as an intermediate in the manufacture of pharmaceuticals like ranitidine and metformin, tramadol and amlodipine. It is used as a precursor of atrazine. It is associated with sodium acetate and used to carry out the Willgerodt- Kindler reaction to prepare amides. Its free base reacts with carbon disulfide to get dimethyldithiocarbamate which is used in rubber vulcanization. It is involved in the synthesis of dimethyl-(1-methyl-pyrrol-2-ylmethyl)-amine by reacting with 1-methyl pyrrole and formaldehyde.

Solubility
Soluble in water, alcohol and chloroform

Notes
Hygroscopic. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 506-59-2
Molecular Formula C2H8ClN
Molecular Weight (g/mol) 81.543
MDL Number MFCD00012477
InChI Key IQDGSYLLQPDQDV-UHFFFAOYSA-N
Synonym dimethylamine hydrochloride, n-methylmethanamine hydrochloride, dimethylammonium chloride, methanamine, n-methyl-, hydrochloride, dimethylamine hcl, dimethylaminehydrochloride, unii-7m4cwb6aok, dimethylamine, hydrochloride, hydrochloric acid dimethylamine, n,n-dimethylamine hydrochloride
PubChem CID 10473
IUPAC Name N-methylmethanamine;hydrochloride
SMILES CNC.Cl

Specifications

Melting Point 168°C to 174°C
Quantity 5000 g
Beilstein 3589311
Sensitivity Hygroscopic
Merck Index 14,3228
Solubility Information Soluble in water,alcohol and chloroform
Formula Weight 81.55
Percent Purity ≥98%
Chemical Name or Material Dimethylamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.