missing translation for 'onlineSavingsMsg'
Learn More

Dimethylamine, 40% w/w aq. soln., Thermo Scientific Chemicals

Product Code. 11347468
Change view
Click to view available options
Quantity:
250 g
1 kg
3 kg
Unit Size:
1kg
250g
3kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347468 3 kg 3kg
11357468 250 g 250g
11367468 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11347468 Supplier Thermo Scientific Alfa Aesar Supplier No. 031458.A5
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Dimethylamine 40% aqueous solution is used as a precursor in the preparation of nitrosodimethylamine. It is also used in paint additives, lubricants, lubricant additives and coating additives. It plays a major role in ion exchange agents and agricultural chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-40-3
Molecular Formula C2H7N
Molecular Weight (g/mol) 45.085
MDL Number MFCD00008288
InChI Key ROSDSFDQCJNGOL-UHFFFAOYSA-N
Synonym dimethylamine, n,n-dimethylamine, methanamine, n-methyl, dimethylamin, dimethyl amine, dimethyl-amine, dimethylamine anhydrous, rcra waste number u092, dimethylamine solution
PubChem CID 674
ChEBI CHEBI:17170
IUPAC Name N-methylmethanamine
SMILES CNC

Specifications

Density 0.890 g/mL
Flash Point 15°C (60°F)
Odor Fish-like
Refractive Index 1.37
Quantity 3 kg
UN Number UN1160
Beilstein 605257
Merck Index 14,3228
Solubility Information Miscible with water and most organic solvents.
Formula Weight 45.08
Concentration or Composition (by Analyte or Components) 40% w/w aq. soln.
Physical Form Liquid
Chemical Name or Material Dimethylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.