Product Code. 30383170
Click to view available options
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
unitSize
30383170
250 g
250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code.
30383170
Supplier
Toronto Research Chemicals
Supplier No.
TRCD460644250G
Please sign in to purchase this item. Need a web account? Register with us today!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Specifications
Chemical Name or Material
Dimethylamine
CAS
124-40-3
Molecular Formula
C2H7N
Quantity
250 g
Synonym
Dimethylamine, N-Methylmethanamine, N,N-Dimethylamine, NSC 8650
InChI Formula
InChI=1S/C2H7N/c1-3-2/h3H,1-2H3
SMILES
CNC
Molecular Weight (g/mol)
45.08
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?