missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Yellow, TRC

Product Code. 30382176
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30382176 1 g
30382975 10 g
30377278 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30382176 Supplier Toronto Research Chemicals Supplier No. TRCD4737901G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-11-7
Molecular Formula C14 H15 N3
Molecular Weight (g/mol) 225.29
Synonym Aniline, N,N-dimethyl-p-phenylazo- (1CI), Benzenamine, N,N-dimethyl-4-(phenylazo)- (9CI), Butter yellow (6CI,7CI), C.I. Solvent Yellow 2 (8CI), N,N-Dimethyl-4-(2-phenyldiazenyl)benzenamine, (4-Dimethylaminophenyl)phenyldiazene, (p-Dimethylaminophenyl)phenyldiazene, 4-(Dimethylamino)azobenzene, 4-(N,N-Dimethylamino)azobenzene, 4-(Phenylazo)-N,N-dimethylaniline, Brilliant Fast Oil Yellow, Brilliant Fast Spirit Yellow, Brilliant Oil Yellow, C.I. 11020, Cerasine Yellow GG, DAB, DAB (carcinogen), DMAB, Dimethyl Yellow, Enial Yellow 2G, Fast Oil Yellow B, Fat Yellow, Fat Yellow A, Fat Yellow AD OO, Fat Yellow ES, Fat Yellow ES Extra, Fat Yellow R, Fat Yellow extra conc, Grasal Brilliant Yellow, Iketon Yellow Extra, Methyl yellow, N,N-Dimethyl-4-(phenylazo)aniline, N,N-Dimethyl-p-(phenylazo)aniline, NSC 6236, Oil Yellow 20, Oil Yellow 2625, Oil Yellow 2G, Oil Yellow BB, Oil Yellow D, Oil Yellow FN, Oil Yellow G, Oil Yellow GG, Oil Yellow GR, Oil Yellow II, Oil Yellow N, Oil Yellow PEL, Oil Yellow S, Oleal Yellow 2G, Organol Yellow ADM, Orient Oil Yellow GG, Petrol Yellow WT, Resinol Yellow GR, Silotras Yellow T 2G, Solvent Yellow 2, Somalia Yellow A, Stear Yellow JB, Sudan Yellow GG, Sudan Yellow GGA, Toyo Oil Yellow G, Waxoline Yellow ADS, Yellow G Soluble in Grease, p-(Dimethylamino)azobenzene
IUPAC Name N,N-dimethyl-4-phenyldiazenylaniline
SMILES CN(C)c1ccc(cc1)N=Nc2ccccc2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C14H15N3/c1-17(2)14-10-8-13(9-11-14)16-15-12-6-4-3-5-7-12/h3-11H,1-2H3
Formula Weight 225.1266
Chemical Name or Material Sudan Yellow
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.