missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Trimethylsilyl Propargylphosphonate, TRC

Product Code. 30383507
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383507 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30383507 Supplier Toronto Research Chemicals Supplier No. TRCD47780050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dimethyl Trimethylsilyl Propargylphosphonate
CAS 228120-29-4
Molecular Formula C8H17O3PSi
Quantity 50 mg
Synonym P-2-Propyn-1-yl-phosphonic Acid Dimethyl Ester, Dimethyl Propargylphosphonate.
InChI Formula InChI=1S/C8H17O3PSi/c1-10-12(9,11-2)7-6-8-13(3,4)5/h7H2,1-5H3
SMILES COP(=O)(CC#C[Si](C)(C)C)OC
IUPAC Name 3-dimethoxyphosphorylprop-1-ynyl(trimethyl)silane
Molecular Weight (g/mol) 220.28
Formula Weight 220.0685
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.