missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl trans-1,4-cyclohexanedicarboxylate, 99+%

Product Code. 10616144 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10616144 25 g Glass Bottle 25g
10087582 1 g Glass Bottle 1g
10551871 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10616144 Supplier Thermo Scientific Acros Supplier No. 240790250
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3399-22-2 | C10H16O4 | 200.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3399-22-2
Molecular Formula C10H16O4
Molecular Weight (g/mol) 200.23
MDL Number MFCD00063917,MFCD00001460,MFCD00216473
InChI Key LNGAGQAGYITKCW-UHFFFAOYSA-N
Synonym dimethyl 1,4-cyclohexanedicarboxylate, dimethyl trans-1,4-cyclohexanedicarboxylate, 1r,4r-dimethyl cyclohexane-1,4-dicarboxylate, cis-dimethyl cyclohexane-1,4-dicarboxylate, dimethyl trans-cyclohexane-1,4-dicarboxylate, 1,4-cyclohexanedicarboxylic acid, dimethyl ester, dimethyl hexahydroterephthalate, trans-dimethyl cyclohexane-1,4-dicarboxylate, 1,4-cyclohexanedicarboxylic acid, dimethyl ester, trans, unii-ys87mds0zv
PubChem CID 7198
IUPAC Name dimethyl cyclohexane-1,4-dicarboxylate
SMILES COC(=O)C1CCC(CC1)C(=O)OC

Specifications

Melting Point 67°C to 70°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass Bottle
Linear Formula CH3OOCC6H10COOCH3
Quantity 25 g
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 200.23
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl trans-1,4-cyclohexanedicarboxylate
Show More Show Less
compliance-icons
Product Identifier
  • Dimethyl trans-1,4-cyclohexanedicarboxylate
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.