missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl trans-1,2-Cyclopropanedicarboxylate, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Dimethyl trans-1,2-Cyclopropanedicarboxylate |
| CAS | 826-35-7 |
| Molecular Formula | C7H10O4 |
| Quantity | 5 g |
| InChI Formula | InChI=1S/C7H10O4/c1-10-6(8)4-3-5(4)7(9)11-2/h4-5H,3H2,1-2H3/t4-,5-/m1/s1 |
| SMILES | COC(=O)[C@@H]1C[C@H]1C(=O)OC |
| IUPAC Name | dimethyl (1R,2R)-cyclopropane-1,2-dicarboxylate |
| Molecular Weight (g/mol) | 158.15 |
| Formula Weight | 158.0579 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction