missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Tetradecanedioate, TRC

Product Code. 30387226
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387226 500 mg 500mg
30375568 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30387226 Supplier Toronto Research Chemicals Supplier No. TRCD460763500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dimethyl Tetradecanedioate
CAS 5024-21-5
Molecular Formula C16 H30 O4
Quantity 500 mg
InChI Formula InChI=1S/C16H30O4/c1-19-15(17)13-11-9-7-5-3-4-6-8-10-12-14-16(18)20-2/h3-14H2,1-2H3
SMILES COC(=O)CCCCCCCCCCCCC(=O)OC
IUPAC Name dimethyl tetradecanedioate
Molecular Weight (g/mol) 286.407
Formula Weight 286.214
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.