missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Terephthalate, Mikromol™

Product Code. 17988814
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17988814 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17988814 Supplier Mikromol Supplier No. MM2520.00
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Dimethyl Terephthalate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Terephthalic acid-bis-methyl ester
CAS 120-61-6
Analyte or Component Names Terephthalic acid-bis-methyl ester
Shipping Condition Room Temperature
Molecular Formula C10 H10 O4
Certifications/Compliance ISO 17034
Synonym 1,4-Benzenedicarboxylic acid dimethyl ester, Terephthalic acid dimethyl ester, DMT, Dimethyl 1,4-benzenedicarboxylate, Dimethyl p-benzenedicarboxylate, Dimethyl p-phthalate, Dimethyl terephthalate, Methyl 4-(carbomethoxy)benzoate, Methyl p-(methoxycarbonyl)benzoate, NSC 3503, 1,4-Benzenedicarboxylic acid, dimethyl ester (9CI), Terephthalic acid, dimethyl ester (6CI,7CI,8CI)
InChI Formula InChI=1S/C10H10O4/c1-13-9(11)7-3-5-8(6-4-7)10(12)14-2/h3-6H,1-2H3
SMILES COC(=O)c1ccc(cc1)C(=O)OC
IUPAC Name dimethyl benzene-1,4-dicarboxylate
Molecular Weight (g/mol) 194.18
Quantity 250 mg
Formula Weight 194.0579 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.