missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Sulfoxide, Tested according to Ph.Eur., Solstice

Product Code. 15675360
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Glass Bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15675360 100 mL Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15675360 Supplier Honeywell-Riedel-de Haen Supplier No. 39291100ML
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
This item is not returnable. View return policy

Tested according to Ph.Eur.

  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-68-5
Density 1.10g/mL
Quantity 100 mL
Explosion Limit 42% (63°F)
Flash Point 87°C (188.6°F)
Ignition Point 573°F
Packaging Glass Bottle
Molecular Formula C2H6OS
Linear Formula (CH3)2SO
Vapor Pressure 0.42mmHg (20°C)
MDL Number MFCD00002089
Beilstein 506008
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
SMILES CS(C)=O
IUPAC Name methanesulfinylmethane
Molecular Weight (g/mol) 78.13
PubChem CID 679
ChEBI CHEBI:28262
Formula Weight 78.13g/mol
Chemical Name or Material Dimethyl sulfoxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.