missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl sulfoxide, Reagent Grade, Solstice

Product Code. 15698480 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
1 gA
4 L
5 L
25 L
230 kg
Unit Size:
1 GAL
1 L
100mL
2.5 L
230kg
25 L
4 L
5 L
500mL
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15698480 4 L 4 L
15667820 100 mL 100mL
15637830 500 mL 500mL
15697820 1 L 1 L
15607830 2.5 L 2.5 L
15647830 5 L 5 L
15627830 25 L 25 L
15687820 1 gA 1 GAL
15617830 230 kg 230kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 15698480 Supplier Honeywell Research Chemicals Supplier No. D58794L

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
SMILES CS(C)=O

Specifications

Density 1.10g/mL
Quantity 4 L
Explosion Limit 42%, 63°F
Flash Point 87°C (188.6°F)
Ignition Point 573°F
Linear Formula (CH3)2SO
Vapor Pressure 0.42 mmHg (20°C)
Beilstein 506008
IUPAC Name methanesulfinylmethane
Formula Weight 78.13g/mol
Percent Purity ≥99.5%
Grade Reagent
Chemical Name or Material Dimethyl sulfoxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.