missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl sulfoxide, puriss. p.a., ACS reagent, Honeywell

Product Code. 15605470
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15605470 100 mL 100mL
15635470 500 mL 500mL
15625470 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15605470 Supplier Honeywell Riedel-de Haën Supplier No. 41640100ML
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
IUPAC Name methylsulfinylmethane
SMILES CS(C)=O

Specifications

Density 1.10g/mL
Quantity 100 mL
Explosion Limit 42%, 63°F
Flash Point 87°C (188.6°F)
Ignition Point 573°F
Linear Formula (CH3)2SO
Vapor Pressure 0.42 mmHg (20°C)
Beilstein 506008
Formula Weight 78.13g/mol
Percent Purity ≥99.9% (GC)
Grade ACS/puriss. p.a.
Chemical Name or Material Dimethyl sulfoxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.