missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl sulfoxide, Puriss. p.a., ≥99.9% (GC), Solstice

Product Code. 15674710
Change view
Click to view available options
Quantity:
6 x 1 L
Packaging:
Glass Bottle
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15674710 6 x 1 L Glass Bottle Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15674710 Supplier Honeywell Riedel-de Haën Supplier No. 349436X1L
Request Bulk or Custom Formats

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss. p.a., dried, ≤0.02% water

  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 16°C to 19°C
CAS 67-68-5
Density 1.10g/mL
Quantity 6 x 1 L
Explosion Limit 42% (63°F)
Flash Point 87°C (188.6°F)
Ignition Point 573°F
Packaging Glass Bottle
Physical Form Liquid
Molecular Formula C2H6OS
Linear Formula (CH3)2SO
Vapor Pressure 0.42mmHg (20°C)
MDL Number MFCD00002089
Beilstein 506008
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
Boiling Point 189°C
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
SMILES CS(C)=O
IUPAC Name methylsulfinylmethane
Molecular Weight (g/mol) 78.13
PubChem CID 679
ChEBI CHEBI:28262
Formula Weight 78.13g/mol
Percent Purity ≥99.9% (GC)
Chemical Name or Material Dimethyl sulfoxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.