missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl sulfoxide, HPLC Grade, 99.9+%

Product Code. 15431107
Change view
Click to view available options
Quantity:
2500 mL
Unit Size:
2500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15431107 2500 mL 2500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15431107 Supplier Thermo Scientific Alfa Aesar Supplier No. 022914.0F
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 67-68-5 | (CH{3}){2}SO

Dimethyl sulfoxide is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 18.4°C
CAS 67-68-5
Quantity 2500 mL
Molecular Formula (CH{3}){2}SO
Boiling Point 189°C
Percent Purity 99.9+%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.