missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl sulfoxide, For ACS analysis, ≥99.9%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
0.5 L
Unit Size:
500mL
Specifications
Specifications
| CAS | 67-68-5 |
| Color | Colorless |
| Density | 1.104g/cm3 at 20°C |
| Quantity | 0.5 L |
| Physical Form | Liquid To Solid |
| Assay Percent Range | ≥99.9% |
| Molecular Formula | C2H6OS |
| MDL Number | MFCD00002089 |
| Synonym | dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide |
| InChI Key | IAZDPXIOMUYVGZ-UHFFFAOYSA-N |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction