missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Sulfoxide, Extra Pure, SLR, Fisher Chemical™

Product Code. p-7072994 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
5 L
25 L
Packaging:
HDPE Bottle
Plastic Container
Unit Size:
2.5 L
25 L
5 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10080110 500 mL HDPE Bottle 500mL
10293800 2.5 L HDPE Bottle 2.5 L
10305800 5 L HDPE Bottle 5 L
10589950 25 L Plastic Container 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10080110 Supplier Fisher Chemical Supplier No. D/4120/PB08
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

C2H6OS, CAS Number-67-68-5, demsodrox, methyl sulfoxide, dimexide, methane, sulfinylbis, dimethyl sulfoxide, dmso, demavet, dimethylsulfoxide, dimethyl sulphoxide, demasorb, 500mL, 189 deg.C, CHEBI:28262, Colorless, 78.13g/mol, IAZDPXIOMUYVGZ-UHFFFAOYSA-N, methylsulfinylmethane, 18.4 deg.C, 2089

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
IUPAC Name methylsulfinylmethane
SMILES CS(C)=O

Specifications

Melting Point 18.4°C
Color Colorless
Quantity 500 mL
Packaging HDPE Bottle
Physical Form Liquid
Vapor Pressure 0.55mbar at 20°C
Viscosity 1.98 mPaS at 25°C
Boiling Point 189°C
Formula Weight 78.13g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.