missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Sulfoxide-d6, TRC
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
Specifications
Specifications
| Chemical Name or Material | Dimethyl Sulfoxide-d6 |
| CAS | 2206-27-1 |
| Molecular Formula | C2 2H6 O S |
| Quantity | 2.5 g |
| Synonym | Sulfinylbismethane-d3, (Methyl Sulfoxide)-d6, Bis(trideuteriomethyl) Sulfoxide, Bis(trideuteromethyl) Sulfoxide, DMSO-d6, Deuterated-DMSO, Di(methyl-d3) Sulfoxide, Dimethyl-d6 Sulfoxide, Hexadeuteriodimethyl Sulfoxide, Perdeuterated Dimethyl Sulfoxide, Perdeuteriodimethyl Sulfoxide, Sulfinylbis(methane-d3) |
| InChI Formula | InChI=1S/C2H6OS/c1-4(2)3/h1-2H3/i1D3,2D3 |
| SMILES | [2H]C([2H])([2H])S(=O)C([2H])([2H])[2H] |
| IUPAC Name | trideuterio(trideuteriomethylsulfinyl)methane |
| Molecular Weight (g/mol) | 84.17 |
| Formula Weight | 84.0516 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction